Learn More
Electron Microscopy Sciences Micromesh 4 Compartment Cassette, Fluo-Pink
SDP
Catalog No. 5031630
Encompass_Preferred
Four compartments have 1676 square openings (0.38 mm) for improved fluid exchange. Made of acetyl polymer, resistant to most histological chemicals. Keeps specimen submerged. Use for labeling machines and tissue processors.
Color: Fluo-Pink
Catalog No. 50-316-30 Supplier Electron Microscopy Sciences Supplier No. 70075FP
Four compartments have 1676 square openings (0.38 mm) for improved fluid exchange. Made of acetyl polymer, resistant to most histological chemicals. Keeps specimen submerged. Use for labeling machines and tissue processors.
Color: Fluo-Pink