Learn More
Electron Microscopy Sciences Micromesh 4 Compartment Cassette, Orange
SDP
Catalog No. 5031627
Encompass_Preferred
Four compartments have 1676 square openings (0.38 mm) for improved fluid exchange. Made of acetyl polymer, resistant to most histological chemicals. Keeps specimen submerged. Use for labeling machines and tissue processors.
Color: Orange
Catalog No. 50-316-27 Supplier Electron Microscopy Sciences Supplier No. 70075O
Four compartments have 1676 square openings (0.38 mm) for improved fluid exchange. Made of acetyl polymer, resistant to most histological chemicals. Keeps specimen submerged. Use for labeling machines and tissue processors.
Color: Orange