Learn More
Electron Microscopy Sciences MicroMesh™ Crystal Mount, 80 µm with 15 µm openings, 25 mm Pin length
SDP
Catalog No. 501898913
Encompass
MicroMesh Mounts are the tool of choice for microcrystal crystallography and diffraction experiments, especially at micro-focus beam-lines. They have been use in de novo protein structure determination from crystals as small as 5um.
They are excellent for rods shaped crystals, and are superior to mount with elliptical apertures, because the mesh provides continuous, gentle support for rods of all sizes.
MicroMesh produce the smallest background scatter of any commercial mount. Their sieve-like action allows easy retrieval of sub 30um crystals.
Mesh: 80 µm
Type: 15 µm openings
Pin length: 25 mm
Catalog No. 50-189-8913 Supplier Electron Microscopy Sciences Supplier No. 6015525
MicroMesh Mounts are the tool of choice for microcrystal crystallography and diffraction experiments, especially at micro-focus beam-lines. They have been use in de novo protein structure determination from crystals as small as 5um.
They are excellent for rods shaped crystals, and are superior to mount with elliptical apertures, because the mesh provides continuous, gentle support for rods of all sizes.
MicroMesh produce the smallest background scatter of any commercial mount. Their sieve-like action allows easy retrieval of sub 30um crystals.
Mesh: 80 µm
Type: 15 µm openings
Pin length: 25 mm