Learn More
Scientific Instrument Services Inc MICROMESH NO12000
Catalog No. NC9565989
Encompass
This item has a minimum qty of 6 per supplier requirements.
Micromesh Abrasives,4x 6 sheet,12000 Grit,green,can be used either wet or dry,have long life, since the crystals will continue to cut effectively until they eventually become dull and lose their sharp cutting edges,do not produce any random
Catalog No. NC9565989 Supplier Scientific Instrument Services Inc Supplier No. MM12000
This item has a minimum qty of 6 per supplier requirements.
Micromesh Abrasives,4x 6 sheet,12000 Grit,green,can be used either wet or dry,have long life, since the crystals will continue to cut effectively until they eventually become dull and lose their sharp cutting edges,do not produce any random