Learn More
StatLab Micromesh Smartload Cassettes
Description
- Taped CS2000
- Small pore size of 0.38 mm allows for efficient fluid exchange and drainage while keeping small biopsies secure
- 45-degree labeling surface, hinged lid, rear tab
Specifications
Specifications
| Color | Pink |
| Product Type | Microbiopsy Cassette |