Learn More
StatLab Micromesh Smartload Cassettes
SDP
Catalog No. 21033426
Change view
Click to view available options
Color:
Green
Pink
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-033-426 Green
21-033-425 Pink
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-033-426 Supplier StatLab Supplier No. M5074T
Add to Cart
Add to Cart
Biopsy cassettes with unique plastic mesh design that eliminates the need for biopsy bags or sponges
  • Taped CS2000
  • Small pore size of 0.38 mm allows for efficient fluid exchange and drainage while keeping small biopsies secure
  • 45-degree labeling surface, hinged lid, rear tab
Specifications
Color Green
Product Type Microbiopsy Cassette