Learn More
Electron Microscopy Sciences Tripod Polisher Micrometer Base
SDP
Catalog No. 50980864
Encompass
Holds the micrometer and has two captive screws that hold the L-Brackets or the Parallel Polishing Mount in position when polishing. For use with Tripod Polisher Model 590.
Catalog No. 50-980-864 Supplier Electron Microscopy Sciences Supplier No. 5011605
Holds the micrometer and has two captive screws that hold the L-Brackets or the Parallel Polishing Mount in position when polishing. For use with Tripod Polisher Model 590.