Learn More
Electron Microscopy Sciences Dual Micrometer Calibration Slide PS52N
SDP
Catalog No. 50367181
Encompass_Preferred
Dual imperial/metric scales calibration slide. 50 mm in 0.1 mm divisions and 2" in 0.005" divisions. In wooden case. Ideal for calibrating optical products with a large field of view, such as stereo microscopes or imaging systems. Negative pattern formed in low reflective chrome for incident light applications. 76 mm x 25 mm.
Catalog No. 50-367-181 Supplier Electron Microscopy Sciences Supplier No. 6804007
Dual imperial/metric scales calibration slide. 50 mm in 0.1 mm divisions and 2" in 0.005" divisions. In wooden case. Ideal for calibrating optical products with a large field of view, such as stereo microscopes or imaging systems. Negative pattern formed in low reflective chrome for incident light applications. 76 mm x 25 mm.