Learn More
MSC Mitutoyo Micrometer Head, 148 Series, 0-0.500 in. Range, 1 ea
Catalog No. 5000515455
Encompass
This small micrometer head is designed for precision positioning and measurement applications. It features a compact design with a 0.375 in. diameter plain stem and a flat spindle face, providing reliable integration into various measuring instruments and fixtures.
  • Spindle range of 0 to 0.500 in.
  • Graduations of 0.001 in. with 0.0001 in. accuracy
  • Equipped with a spindle lock for secure positioning
  • Zero-adjustable at any position by loosening the setscrew
  • Hard-chrome plated thimble with laser-etched scales for clear readability
  • Lapped spindle face constructed of SKS3 steel
Catalog No. 50-005-15455 Supplier MSC Supplier No. 97237143
This small micrometer head is designed for precision positioning and measurement applications. It features a compact design with a 0.375 in. diameter plain stem and a flat spindle face, providing reliable integration into various measuring instruments and fixtures.
  • Spindle range of 0 to 0.500 in.
  • Graduations of 0.001 in. with 0.0001 in. accuracy
  • Equipped with a spindle lock for secure positioning
  • Zero-adjustable at any position by loosening the setscrew
  • Hard-chrome plated thimble with laser-etched scales for clear readability
  • Lapped spindle face constructed of SKS3 steel