Learn More
ZORO TOOLS MICROMETER INSIDE 5/8 TO 3/4"
Catalog No. NC2195078
Encompass
STARRETT Micrometer Inside 5/8 to 3/4" Range. item: Bore Gage Micrometer. Resolution: 0.00025".
This item is not returnable. View return policy
Catalog No. NC2195078 Supplier ZORO TOOLS Supplier No. G0583007
STARRETT Micrometer Inside 5/8 to 3/4" Range. item: Bore Gage Micrometer. Resolution: 0.00025".