Learn More
Electron Microscopy Sciences Micrometer, Model A
SDP
Catalog No. 50996486
Encompass_Preferred
Metric and English scale micrometers with friction stop; comes in a padded plastic case. Calibrated to read 0 to 25mm, in 0.01mm divisions.
Catalog No. 50-996-486 Supplier Electron Microscopy Sciences Supplier No. 62060
Metric and English scale micrometers with friction stop; comes in a padded plastic case. Calibrated to read 0 to 25mm, in 0.01mm divisions.