Learn More
Electron Microscopy Sciences Micrometer, Model B
SDP
Catalog No. 50996487
Encompass_Preferred
Metric and English scale micrometers with friction stop; comes in a padded plastic case. Calibrated to read 0 to 1", in 0.001" divisions.
Catalog No. 50-996-487 Supplier Electron Microscopy Sciences Supplier No. 62061
Metric and English scale micrometers with friction stop; comes in a padded plastic case. Calibrated to read 0 to 1", in 0.001" divisions.