Learn More
Electron Microscopy Sciences Micrometer Scale Reflected light
SDP
Catalog No. 501901567
Encompass_Preferred
Reflected light calibration slide with 0.1" length scale subdivided into 0.001" divisions
Catalog No. 50-190-1567 Supplier Electron Microscopy Sciences Supplier No. 68047PS4R
Reflected light calibration slide with 0.1" length scale subdivided into 0.001" divisions