Learn More
Glass Expansion MicroMist DC Nebulizer 0.05mL/min
SDP
Catalog No. 5000556403
Encompass_Preferred
MicroMist DC Nebulizer 0.05mL/min Sourced from Glass Expansion
Catalog No. 50-005-56403 Supplier Glass Expansion Supplier No. A231UM005
MicroMist DC Nebulizer 0.05mL/min Sourced from Glass Expansion