Learn More
Glass Expansion MicroMist DC Nebulizer 0.05mL/min, 1.6mm OD
SDP
Catalog No. 502703012
Encompass_Preferred
MicroMist DC Nebulizer 0.05 mL/min, includes UniFit Connector with 0.25mm ID x 1.6mm OD x 700mm long tubing (1 pc.) and DC Gas Fitting Type 13/31 with ratchet connector (1 pc.). Compatible with Thermo PRO Duo, PRO Radial, 6000/7000 Duo & Radial Series ICP-OES.Optimum nebulizer gas flow rate is 0.7L/min and sample flow rate is 0.05mL/min. Recommended not to exceed optimum 0.05mL/min sample flow rate; however, lower gas and sample flow rates are OK. The backpressure should read ~40 psi at 0.7L/min gas flow rate. Can tolerate up to 15% TDS. MicroMist is the standard configuration for many ICP manufacturers due to its excellent sensitivity and precision at low flow rates.Replacement parts include P/Ns NFT-025 and 501907289.Recommend maintenance tools are Fluka RBS25 (P/N NC9000117) and Eluo Nebulizer Cleaning Tool (P/N NC9462559). When not in use, best to store in the provided hard plastic case.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3012 Supplier Glass Expansion Supplier No. A3107UM005
MicroMist DC Nebulizer 0.05 mL/min, includes UniFit Connector with 0.25mm ID x 1.6mm OD x 700mm long tubing (1 pc.) and DC Gas Fitting Type 13/31 with ratchet connector (1 pc.). Compatible with Thermo PRO Duo, PRO Radial, 6000/7000 Duo & Radial Series ICP-OES.Optimum nebulizer gas flow rate is 0.7L/min and sample flow rate is 0.05mL/min. Recommended not to exceed optimum 0.05mL/min sample flow rate; however, lower gas and sample flow rates are OK. The backpressure should read ~40 psi at 0.7L/min gas flow rate. Can tolerate up to 15% TDS. MicroMist is the standard configuration for many ICP manufacturers due to its excellent sensitivity and precision at low flow rates.Replacement parts include P/Ns NFT-025 and 501907289.Recommend maintenance tools are Fluka RBS25 (P/N NC9000117) and Eluo Nebulizer Cleaning Tool (P/N NC9462559). When not in use, best to store in the provided hard plastic case.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001