Learn More
Glass Expansion MicroMist DC Nebulizer 0.2mL/min, Includes NFT-16-50 UniFit Connector with 0.50mm ID x 1.6mm OD x 700mm long tubing (1 piece), and 70-803-1049 Gas Fitting with Ratchet Nebulizer connector
SDP
Catalog No. 501907456
Encompass_Preferred
MicroMist DC Nebulizer 0.2 mL/min, includes UniFit Connector with 0.50mm ID x 1.6mm OD x 700mm long tubing (1 pc.) and DC Gas Fitting Type 22 with ratchet connector (1 pc.). Compatible with Perkin Elmer Elan/NexION 300/350.Optimum nebulizer gas flow rate is 1.0L/min and sample flow rate is 0.2mL/min. Recommended not to exceed optimum 0.2mL/min sample flow rate; however, lower gas and sample flow rates are OK. The backpressure should read ~40 psi at 1.0L/min gas flow rate. Can tolerate up to 15% TDS. MicroMist is the standard configuration for many ICP manufacturers due to its excellent sensitivity and precision at low flow rates. Replacement parts include P/Ns 502702959 and 502703069.Recommend maintenance tools are Fluka RBS25 (P/N NC9000117) and Eluo Nebulizer Cleaning Tool (P/N NC9462559). When not in use, best to store in the provided hard plastic case.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7456 Supplier Glass Expansion Supplier No. A221UM02
MicroMist DC Nebulizer 0.2 mL/min, includes UniFit Connector with 0.50mm ID x 1.6mm OD x 700mm long tubing (1 pc.) and DC Gas Fitting Type 22 with ratchet connector (1 pc.). Compatible with Perkin Elmer Elan/NexION 300/350.Optimum nebulizer gas flow rate is 1.0L/min and sample flow rate is 0.2mL/min. Recommended not to exceed optimum 0.2mL/min sample flow rate; however, lower gas and sample flow rates are OK. The backpressure should read ~40 psi at 1.0L/min gas flow rate. Can tolerate up to 15% TDS. MicroMist is the standard configuration for many ICP manufacturers due to its excellent sensitivity and precision at low flow rates. Replacement parts include P/Ns 502702959 and 502703069.Recommend maintenance tools are Fluka RBS25 (P/N NC9000117) and Eluo Nebulizer Cleaning Tool (P/N NC9462559). When not in use, best to store in the provided hard plastic case.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001