Learn More
Glass Expansion MicroMist HE U-Series Nebulizer 0.2mL/min, Used for single-cell analysis of biological materials. Compatible with Helios, CyTOF and CyTOF2. Also used in any high efficiency applications, such as single particle and nano particle studies.
SDP
Catalog No. 502702995
Encompass_Preferred
MicroMist HE U-Series Nebulizer 0.2mL/min, Used for single-cell analysis of biological materials. Compatible with Helios, CyTOF and CyTOF2. Also used in any high efficiency applications, such as single particle and nano particle studies.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2995 Supplier Glass Expansion Supplier No. C211UFT02
MicroMist HE U-Series Nebulizer 0.2mL/min, Used for single-cell analysis of biological materials. Compatible with Helios, CyTOF and CyTOF2. Also used in any high efficiency applications, such as single particle and nano particle studies.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001