Learn More
Glass Expansion MicroMist Nebulizer 0.2mL/min with EzyFit & EzyLok
SDP
Catalog No. 5000555644
Encompass_Preferred
MicroMist Nebulizer 0.2mL/min with EzyFit & EzyLok Sourced from Glass Expansion
Catalog No. 50-005-55644 Supplier Glass Expansion Supplier No. AR351FM02LE
MicroMist Nebulizer 0.2mL/min with EzyFit & EzyLok Sourced from Glass Expansion