Learn More
Glass Expansion MicroMist U-Series Nebulizer 0.1mL/min, Includes NFT-16-25 UniFit Connector with 0.25mm ID x 1.6mm OD x 700mm long tubing (1 piece), and EL-1 EzyLok Connector Kit
SDP
Catalog No. 501907523
Encompass_Preferred
MicroMist Nebulizer 0.1 mL/min, includes UniFit Connector with 0.25mm ID x 1.6mm OD x 700mm long tubing (1 pc.) and EzyLok Connector (1 pc.).Optimum nebulizer gas flow rate is 0.7L/min and sample flow rate is 0.1mL/min. Recommended not to exceed optimum 0.1 mL/min sample flow rate; however, lower gas and sample flow rates are OK. The backpressure should read ~40psi at 0.7L/min gas flow rate. Can tolerate up to 15% TDS. MicroMist is the standard configuration for many ICP-MS manufacturers due to its excellent sensitivity and precision.Replacement parts include P/Ns NFT-16-25 and 502702994.Recommend nebulizer maintenance tools, Fluka RBS25 (P/N NC9000117) and Eluo Nebulizer Cleaning Tool (P/N NC9462559). When nebulizer is not in use, best to store in the provided hard plastic case.Consider upgrading to our new Direct Connect (DC) MicroMist to take advantage of the cost-effective design improvements.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7523 Supplier Glass Expansion Supplier No. ARG07UM01
MicroMist Nebulizer 0.1 mL/min, includes UniFit Connector with 0.25mm ID x 1.6mm OD x 700mm long tubing (1 pc.) and EzyLok Connector (1 pc.).Optimum nebulizer gas flow rate is 0.7L/min and sample flow rate is 0.1mL/min. Recommended not to exceed optimum 0.1 mL/min sample flow rate; however, lower gas and sample flow rates are OK. The backpressure should read ~40psi at 0.7L/min gas flow rate. Can tolerate up to 15% TDS. MicroMist is the standard configuration for many ICP-MS manufacturers due to its excellent sensitivity and precision.Replacement parts include P/Ns NFT-16-25 and 502702994.Recommend nebulizer maintenance tools, Fluka RBS25 (P/N NC9000117) and Eluo Nebulizer Cleaning Tool (P/N NC9462559). When nebulizer is not in use, best to store in the provided hard plastic case.Consider upgrading to our new Direct Connect (DC) MicroMist to take advantage of the cost-effective design improvements.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001