Learn More
Profoldin MicroMolar Histidine Assay Kit
Catalog No. NC1709403
Encompass
This product includes 750 ul 10 x C53 Dye and 50 ul of 10 mM Histidine. It is for measurement of 200 samples using 96-well plates. Cuvettes may also be used for measurements.
Catalog No. NC1709403 Supplier Profoldin Supplier No. HIS200
This product includes 750 ul 10 x C53 Dye and 50 ul of 10 mM Histidine. It is for measurement of 200 samples using 96-well plates. Cuvettes may also be used for measurements.