Learn More
Fine Science Tools Micro-Mosquito Hemostats, Stainless Steel, 12 cm
SDP
Catalog No. NC9934326
Encompass
Tip Shape: Straight/ Tips: Serrated/ Tip Width: 1.3mm/ Clamping Length: 20mm/ Alloy / Material: Stainless Steel/ Length: 12cm
Catalog No. NC9934326 Supplier Fine Science Tools Supplier No. 1301012
Tip Shape: Straight/ Tips: Serrated/ Tip Width: 1.3mm/ Clamping Length: 20mm/ Alloy / Material: Stainless Steel/ Length: 12cm