Learn More
Leica Biosystems MICROMOUNT MOUNTING MEDIA 1PT
Catalog No. NC1109222
Encompass_Preferred
Micromount is a rapid-drying, xylene-based mounting medium. An antioxidant is added to inhibit stain fading and prevent the formation of annual rings. Micromount sets up quickly, allowing slide preparations to be handled immediately after mounting. Micromount is soluble in xylene, toluene, and most xylene substitutes.
Catalog No. NC1109222 Supplier Leica Biosystems Supplier No. 3801731
Micromount is a rapid-drying, xylene-based mounting medium. An antioxidant is added to inhibit stain fading and prevent the formation of annual rings. Micromount sets up quickly, allowing slide preparations to be handled immediately after mounting. Micromount is soluble in xylene, toluene, and most xylene substitutes.