Learn More
Electron Microscopy Sciences MicroMounts™ Crystal Tool with 20 µm Aperture and SPINE pin
SDP
Catalog No. 509053339
Encompass
MicroMounts are the high performance tools for manipulating and mounting protein crystals, virus crystals, and small molecule/inorganic crystals, and other small, fragile samples. They are ideal for both conventional and high-throughput X-ray crystallography applications.
Each consist of a thin (7.5um thick) micro-fabricated polyimide film, attached to a solid non-magnetic stainless steel pin. Film is wrapped around the steel pin and it provides excellent stiffness and convenient scoop-like action in retrieving and handling crystals.
Aperture: SPINE pin
Catalog No. 50-905-3339 Supplier Electron Microscopy Sciences Supplier No. 60116S
MicroMounts are the high performance tools for manipulating and mounting protein crystals, virus crystals, and small molecule/inorganic crystals, and other small, fragile samples. They are ideal for both conventional and high-throughput X-ray crystallography applications.
Each consist of a thin (7.5um thick) micro-fabricated polyimide film, attached to a solid non-magnetic stainless steel pin. Film is wrapped around the steel pin and it provides excellent stiffness and convenient scoop-like action in retrieving and handling crystals.
Aperture: SPINE pin