Learn More
Electron Microscopy Sciences MicroMounts™ Crystal Tool with Assortment A1, 25 mm pin
SDP
Catalog No. 509053319
Encompass
MicroMounts are the high performance tools for manipulating and mounting protein crystals, virus crystals, and small molecule/inorganic crystals, and other small, fragile samples. They are ideal for both conventional and high-throughput X-ray crystallography applications.
Each consist of a thin (7.5um thick) micro-fabricated polyimide film, attached to a solid non-magnetic stainless steel pin. Film is wrapped around the steel pin and it provides excellent stiffness and convenient scoop-like action in retrieving and handling crystals. Includes: 10, 20, 30 and 40 um apertures.
Aperture: 25 mm pin
Catalog No. 50-905-3319 Supplier Electron Microscopy Sciences Supplier No. 6011025
MicroMounts are the high performance tools for manipulating and mounting protein crystals, virus crystals, and small molecule/inorganic crystals, and other small, fragile samples. They are ideal for both conventional and high-throughput X-ray crystallography applications.
Each consist of a thin (7.5um thick) micro-fabricated polyimide film, attached to a solid non-magnetic stainless steel pin. Film is wrapped around the steel pin and it provides excellent stiffness and convenient scoop-like action in retrieving and handling crystals. Includes: 10, 20, 30 and 40 um apertures.
Aperture: 25 mm pin