Learn More
Cayman Chemical Micronomicin (sulfate), Purity: ≥95% (contains up to 40% gentamicin C1a), CAS: 66803-19-8, 500 mg
SDP
Catalog No. 502940231
Encompass_Preferred
An aminoglycoside antibiotic; active against S. aureus, B. subtilis, B. cereus, E. coli, and K. pneumoniae (MICs = 0.001-8.3 μg/ml); inactive against C. albicans and A. niger (MICs = >10 μg/ml)
Catalog No. 50-294-0231 Supplier Cayman Chemical Supplier No. 27951500 MG
An aminoglycoside antibiotic; active against S. aureus, B. subtilis, B. cereus, E. coli, and K. pneumoniae (MICs = 0.001-8.3 μg/ml); inactive against C. albicans and A. niger (MICs = >10 μg/ml)