Learn More
Medchemexpress LLC HY-108307 50mg Medchemexpress, Micronomicin (sulfate) CAS:66803-19-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10830750MG
Encompass_Preferred
Medchemexpress, HY-108307 50mg Micronomicin (sulfate) CAS:66803-19-8 Micronomicin sulfate (Gentamicin C2b sulfate) is an aminoglycoside antibiotic isolated from Micromonospora. Micronomicin sulfate is a broad-spectrum antibiotic close to the gentamicin-type antibiotics, exhibits a high activity against Pseudomonas, Proteus, Klebsiella pneumoniae, Serratia, etc (MIC=0.001-8.3 μg/ml). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0737