Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Micronomicin sulfate, 590 ^mg/mg

Catalog No. AAJ6607209
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6607209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6607209 Supplier Thermo Scientific Chemicals Supplier No. J6607209
Add to Cart

CAS: 66803-19-8 | C20H43N5O11S | 561.65 g/mol

An aminoglycoside antibiotic commonly used against Gram-negative bacteria

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • An aminoglycoside antibiotic commonly used against gram negative bacteria
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66803-19-8
Molecular Formula C20H43N5O11S
Molecular Weight (g/mol) 561.65
MDL Number MFCD00941451
InChI Key MQDGQSCLOYLSEK-SCFBDNQUSA-N
PubChem CID 171808
IUPAC Name (2R,3R,4R,5R)-2-{[(1S,2S,3R,4S,6R)-4,6-diamino-3-{[(2R,3R,6S)-3-amino-6-[(methylamino)methyl]oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}-5-methyl-4-(methylamino)oxane-3,5-diol; sulfuric acid
SMILES OS(O)(=O)=O.CNC[C@@H]1CC[C@@H](N)[C@@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O[C@H]3OC[C@](C)(O)[C@H](NC)[C@H]3O)[C@H]2O)O1

Specifications

Type Micronomicin Sulfate
Physical Form Powder
Color White to Yellow
Quantity 10 g
Flash Point >110°C (230°F)
Boiling Point 667°C
Solubility Information Freely soluble in water
Formula Weight 561.65
Chemical Name or Material Micronomicin sulfate, 590μg/mg
RTECSNumber WK2196000
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.