Learn More
Micronova™ Double-Coated Cleanroom Tape
SDP
Catalog No. 19058391 Shop All Micronova Manufacturing Products
Change view
Click to view available options
0.5 in. x 36 yd.
1 in. x 36 yd.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-058-391 0.5 in. x 36 yd.
19-058-390 1 in. x 36 yd.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-058-391 Supplier Micronova™ Supplier No. 12TRDC
Made of PVC film coated with a long-lasting acrylic adhesive on each side, this linerless double-coated film tape is designed for cleanroom applications.
•    Differential adhesive eliminates the need to remove and discard liner material, so there is less waste
•    Adhesive bonds with varied materials, including metals, plastics, and glass
•    Extremely resistant to solvents and UV
Order Info
Wound on a plastic core in 36 yd. (33m) rolls
Compliance
Meets or exceeds UU-T-91E, Type 1