Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Micronova™ Stainless Steel Gel Cleaner
GSA_VASDP

Catalog No. 19305710 Shop All Micronova Manufacturing Products
Change view
Click to view available options
Volume (English):
16 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19-305-710 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-305-710 Supplier Micronova™ Supplier No. SC1P
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stainless steel gel cleanser spray, with advanced no-drip technology, for removing rust, oxidation and residue build-up on stainless steel, metal and many other hard surfaces. Available in a 16 oz. spray bottle.

  • Clings and glides over most surfaces with minimum effort on contact
  • Apply into seams and crevices, when dealing with rust, oxidation and residue build-up to pre-treat before wiping
  • Nonatomized — will not affect air quality
  • Phosphorus-free
  • Biodegradable surfactants
  • No VOCs
  • NSF A7 rating

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Gel Detergent
Color-coded No
Form Gel
Handle Type None
Sterility Non-sterile
Appearance Clear
Color Clear
Content And Storage Store at ambient temperature. Avoid direct sunlight. Protect from freezing.
For Use With (Application) Gel Cleaner for Stainless Surface
Volume (English) 16 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.