Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Micronova™ IPA Resistant LabMarker
BIOPHARMAGSA_VASDP

Catalog No. 19142679 Shop All Micronova Manufacturing Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-142-679 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-142-679 Supplier Micronova™ Supplier No. PEN40
  • PRODUCT PROMOTION
    $189.00
    $53.61 / Pack of 10

    Save $135.39

    72% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A fine tipped permanent marker with alcohol-resistant ink that marks on cold or wet surfaces, plastic bags and disposable lab-ware.

With permanent, quick drying black ink that is resistant to IPA

The Micronova IPA Resistant LabMarker features permanent, quick drying, alcohol-resistant ink that marks on a range of surfaces and will not smear on reusable glass, metal or porcelain, but can be scrubbed off. Available gamma irradiated for aseptic areas.

  • Fine-tip, black permanent marker
  • Quick drying, alcohol-resistant ink
  • Marks on cold or wet surfaces, plastic bags, and disposable lab-ware
  • Smear-resistant on glass, metal and porcelain, but can be scrubbed off
  • Available gamma irradiated for aseptic areas

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Cleanroom Pen
Tip Style Fine
Color Black
Autoclavable Not Autoclavable
Sterility Non-sterile
Material (Barrel) High-Density Polypropylene
Disposable Reusable
For Use With (Application) Cleanroom and Aseptic Environment Documentation and Labeling
No. per Pack 10
Quantity Ea.
Capacity (Metric) 2 mL
Length (English) 5.25 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.