Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Micronova™ Quick Disconnect MicroMop™ Adapter
SDP

Catalog No. 19035363A Shop All Micronova Manufacturing Products
Change view
Click to view available options
Autoclavable:
Autoclavable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Autoclavable
19-035-363A Autoclavable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-035-363A Supplier Micronova™ Supplier No. QDMH1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Polypropylene clamp adapter to hold MicroMop™ and PolyGen series of floor mops. Attaches to SSU series of handles.

  • QDMH-1 adapter; connects to stainless-steel SSU series handles
  • Orange polypropylene
  • Lightweight and durable
  • Features a stainless steel, quick-disconnect handle attachment
  • Autoclavable

Note: This is only an adapter. Handle must be purchased separately. (Compatible handles include the SSU series.)

Specifications

Product Type Mop Adapter
Color Orange
Length (English) 16 in.
Sterility Non-sterile
Length (Metric) 41 cm
Autoclavable Autoclavable
Disposable Reusable
Care Instructions Store at Ambient Temperature. Rinse to remove Disinfectant before Autoclave or Storage
Description Adapter to hold MicroMop and PolyGen™ floor mops. Attaches to SSU series of handles,.
For Use With (Application) For Cleaning Large Surface Areas Quickly and Efficiently
Color-coded No
Material Electropolished Stainless Steel (304), Polyethylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.