Learn More
Micronova™ PowerHead™ Handle
SDP
Catalog No. 19041620 Shop All Micronova Manufacturing Products
Change view
Click to view available options
Autoclavable:
Autoclavable
Catalog No. Autoclavable
19-041-620 Autoclavable
1 options
Catalog No. 19-041-620 Supplier Micronova™ Supplier No. PHB10
Efficiently wrings all PowerHead mop heads
PowerHead Handle
  • Has built-in quick-disconnect fitting that allows it to be used with single-length or extendible Micronova universal handles
  • For use in ISO Classes 3 to 8 (Class 1 to Class 100,000) cleanroom
Specifications
Product Type Mop Adapter
Autoclavable Autoclavable
Width (English) 10 in.
For Use With (Equipment) Retro Fitting Commercial Mop Handles to PowerHead System
For Use With (Application) For ISO Class 3 to 8 Cleanrooms
Material Stainless Steel