Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Micronova™ Micronova™ Slim-T™ Isolator Tool Cover
GREENER_CHOICESDP

Catalog No. 19142660 Shop All Micronova Manufacturing Products
Catalog No. 19-142-660 Supplier Micronova™ Supplier No. ITC119NSBB
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Micronova™ Slim-T™ Isolator Tool Cover is an easy on and off small mop cover designed for efficient cleaning and disinfecting isolators, hoods, RABS and small equipment in clean and aseptic areas.

  • Modular and extendable handles fit in small pass throughs, yet expand reach once open
  • Ergonomic method for cleaning small equipment and small spaces
  • 360 degree swivel
  • A variety of fabrics increase effectiveness in differing environments
  • Smooth 100% polyester, NovaPoly covers are ideal for surface disinfection
  • Textured polyester design combines efficient dispersal of disinfectant with effective cleaning
  • MegaTex pad on polyester cover allows increased absorbency and minimal abrasion.
  • Envelope design allows for simple attachment without clips, snaps or fastenings, and easy removal
  • Compatible with alcohol, strong disinfectants and steam sterilization

Specifications

Product Type Cleanroom Mop
Head Type Small Flat Head
Color White
Autoclavable Autoclavable
For Use With (Equipment) QPIT and QDIT Series Isolator Tools
Description PolySorb Isolator Tool Cover, 9 in., Bulk Pack
Color-coded No
Material Textured Polyester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.