Description
- Vinyl tape features nontransferring adhesive and a 3 in. (7.6cm) poly core.
- Resists most acids, alkalis, and bacteria
- Tape easily conforms to curved and irregular surfaces
- Matte finish can be written on with a cleanroom pen
Order Info
- Available in a variety of colors and sizes for coding, identification, sealing, labeling, or aisle marking.
- Roll length, 36 yards (32.9m)
Specifications
Specifications
| Color | White |
| Dimensions (L x W) | 36 yd x 2 in. |