Learn More
World Precision Instrument Microsensor Adapter Cable
SDP
Supplier:  World Precision Instrument 91580
Encompass
  • For use with Nitric Oxide, Hydrogen Peroxide, Hydrogen Sulfide micro sensors for use with the TBR4100 or TBR1025 free radical analyzers.
  • Catalog No. 50-252-1217