Learn More
StatLab Microorganism Identification and Confirmation Discs
SDP
Catalog No. 22130507
Change view
Click to view available options
Quantity:
50/Pk.
Catalog No. Quantity
22-130-507 50/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 22-130-507 Supplier StatLab Supplier No. 3981538
Add to Cart
Add to Cart
This item is not returnable. View return policy
Used for the identification and confirmation of microorganisms
  • Easy-to-use with dependable, easy-to-read results
  • Based on rapid screening methods with a reaction time of less than 30 minutes
  • 50 discs/pkg
Specifications
Format Paper Disc
Product Type Microbial Identification Disc
Quantity 50/Pk.