Learn More
Electron Microscopy Sciences MicroPen-tek Pen with Flat Round Tip
SDP
Catalog No. 501931032
Encompass_Preferred
The MicroPen-tek safely and effectively cleans micro camera lenses. With its specially designed flat and round micro-tip, it applies the invisible carbon compound to places its other cleaning pens can't reach. Retractable brush removes dust and the unique invisible carbon compound outperforms all other cleaning systems.
Catalog No. 50-193-1032 Supplier Electron Microscopy Sciences Supplier No. 7832589
The MicroPen-tek safely and effectively cleans micro camera lenses. With its specially designed flat and round micro-tip, it applies the invisible carbon compound to places its other cleaning pens can't reach. Retractable brush removes dust and the unique invisible carbon compound outperforms all other cleaning systems.