Learn More
Research Products International Corp Micro-Pestles with 1.5ml Micro-Tubes, Disposable, 100 per Case
SDP
Catalog No. 50212457
Encompass
These autoclavable/disposable plastic pestles are excellent for re-suspending bacterial cells, precipitates of nucleic acids or proteins and pellets of other materials. Pestle supplied in green color.
Catalog No. 50-212-457 Supplier Research Products International Corp Supplier No. 199222
These autoclavable/disposable plastic pestles are excellent for re-suspending bacterial cells, precipitates of nucleic acids or proteins and pellets of other materials. Pestle supplied in green color.