Learn More
World Precision Instrument Micropipette Tips for Nanoliter 2020/2010
SDP
Catalog No. 50821958
Encompass
Eliminate the cost and trouble of making your own micropipettes
WPI can quickly supply your need for pre-pulled glass micropipettes for use with Nanoliter Injectors. These tips are sold in packages of 10, in vacuum packages. With a tip inside diameter of 10µm and a shank outside diameter of 1.14 mm. These MicroTips are made with glass capillaries (#504949), which is fire polished, so the shank is fire polished and the tip is not. They have no filament. Each one is quality tested.
Catalog No. 50-821-958 Supplier World Precision Instrument Supplier No. TIP10XV119
Eliminate the cost and trouble of making your own micropipettes
WPI can quickly supply your need for pre-pulled glass micropipettes for use with Nanoliter Injectors. These tips are sold in packages of 10, in vacuum packages. With a tip inside diameter of 10µm and a shank outside diameter of 1.14 mm. These MicroTips are made with glass capillaries (#504949), which is fire polished, so the shank is fire polished and the tip is not. They have no filament. Each one is quality tested.