Learn More
World Precision Instrument Pre-Pulled Glass Pipettes, Luer-TIP01TW1F-L
SDP
Catalog No. 50821945
Encompass
WPI can quickly supply your need for consistently sized, pre-pulled, glass, calibrated micropipettes. Tip diameters (ID) range from 0.1 to 10 µm. Silanized Tips (Luer Shank) are available. Silanization waterproofs the glass to retard water when inserting into cell. This will not let the outside fluid run down the glass pipettes and get inside so easily.These glass needles are fabricated from Schott Duran borosilicate glass.
Catalog No. 50-821-945 Supplier World Precision Instrument Supplier No. TIP01TW1FL
WPI can quickly supply your need for consistently sized, pre-pulled, glass, calibrated micropipettes. Tip diameters (ID) range from 0.1 to 10 µm. Silanized Tips (Luer Shank) are available. Silanization waterproofs the glass to retard water when inserting into cell. This will not let the outside fluid run down the glass pipettes and get inside so easily.These glass needles are fabricated from Schott Duran borosilicate glass.