Learn More
World Precision Instrument Pre-Pulled Glass Pipettes, Silanized-TIP5TW1LS01
SDP
Catalog No. 50821973
Encompass
WPI can quickly supply your need for consistently sized pre-pulled glass micropipettes. Tip diameters (ID) range from 0.1 to 10 µm. Silanized Tips (Luer Shank) are available. Silanization waterproofs the glass to retard water when inserting into cell. This will not let the outside fluid run down the pipette and get inside so easily.These glass needles are fabricated from Schott Duran borosilicate glass.
Catalog No. 50-821-973 Supplier World Precision Instrument Supplier No. TIP5TW1LS01
WPI can quickly supply your need for consistently sized pre-pulled glass micropipettes. Tip diameters (ID) range from 0.1 to 10 µm. Silanized Tips (Luer Shank) are available. Silanization waterproofs the glass to retard water when inserting into cell. This will not let the outside fluid run down the pipette and get inside so easily.These glass needles are fabricated from Schott Duran borosilicate glass.