Learn More
Miles Scientific CALIBRATED MICROPIPETTES - 1-5UL (PKG 250)
SDP
Catalog No. NC0127687
Encompass_Preferred
Economical capillary micropipettes with fire polished tips and calibration marks at 1,2,3,4, and 5 microliters. Accuracy is within 1%. Total length of each capillary is five inches for easy handling. A 15” aspirator tube with mouthpiece and 250 micropipettes are included with each package. 
Catalog No. NC0127687 Supplier Miles Scientific Supplier No. A2040
Economical capillary micropipettes with fire polished tips and calibration marks at 1,2,3,4, and 5 microliters. Accuracy is within 1%. Total length of each capillary is five inches for easy handling. A 15” aspirator tube with mouthpiece and 250 micropipettes are included with each package. 