Learn More
RWD LIFE SCIENCE INC MICROPIPETTE PULLER
SDP
Catalog No. NC3351675
Encompass_Preferred
MP-1000 Micropipette Puller is a horizontal motorized micropipette puller, designed to solve the microelectrode problems in patch clamp and microinjection experiments. MP-1000 integrates stability, intelligence and humanization, which can meet the requirements of microelectrode-related experiments and enhance user experience at the same time. In summary, RWD MP-1000 Micropipette Puller is an excellent choice for micropipette related experiments.
Catalog No. NC3351675 Supplier RWD LIFE SCIENCE INC Supplier No. MP1000
Add to Cart
Add to Cart
Request a Quote
MP-1000 Micropipette Puller is a horizontal motorized micropipette puller, designed to solve the microelectrode problems in patch clamp and microinjection experiments. MP-1000 integrates stability, intelligence and humanization, which can meet the requirements of microelectrode-related experiments and enhance user experience at the same time. In summary, RWD MP-1000 Micropipette Puller is an excellent choice for micropipette related experiments.