Learn More
Chemglass Life Sciences Deep 24-Well Microplate, 10.4 mL Wells, V-Bottom, Square Top, Sterile
SDP
Catalog No. 502733178
Encompass_Preferred
Deep 24-well polypropylene microplate with 10.4 mL capacity per well and V-bottoms, supplied sterile for sample storage, assays, and tissue culture workflows. The nested plate design facilitates handling and compact storage.
  • Deep 24-well format with 10.4 mL capacity per well.
  • V-bottom wells for efficient sample recovery.
  • Square well top for stacking and compatibility with automation.
  • Made from USP VI polypropylene for chemical compatibility.
  • Supplied sterile and nested for convenient handling and storage.
  • Packaged CS5 (case of 5) for laboratory supply management.
Catalog No. 50-273-3178 Supplier Chemglass Life Sciences Supplier No. CLS3506S242
Deep 24-well polypropylene microplate with 10.4 mL capacity per well and V-bottoms, supplied sterile for sample storage, assays, and tissue culture workflows. The nested plate design facilitates handling and compact storage.
  • Deep 24-well format with 10.4 mL capacity per well.
  • V-bottom wells for efficient sample recovery.
  • Square well top for stacking and compatibility with automation.
  • Made from USP VI polypropylene for chemical compatibility.
  • Supplied sterile and nested for convenient handling and storage.
  • Packaged CS5 (case of 5) for laboratory supply management.