Learn More
Chemglass Life Sciences Microplate, 24-Well Deep Well, 16 mL V-Bottom (Sterile, Nest-Pack, Includes Comb)
SDP
Catalog No. 502733176
Encompass_Preferred
Chemglass deep-well 24-well microplate with 16 mL V-bottom wells, provided sterile and nest-packed for laboratory sample processing and storage. Includes a 24-well plastic comb and is suitable for automated workflows.
  • 24 wells
  • 16 mL nominal volume per well
  • V-bottom well geometry
  • Square well top for secure sealing
  • Manufactured from USP Class VI polypropylene
  • Sterile, nest-packed format
  • Includes a 24-well plastic comb
  • Suitable for storage, extraction, and automated processing
Catalog No. 50-273-3176 Supplier Chemglass Life Sciences Supplier No. CLS3506S245
Chemglass deep-well 24-well microplate with 16 mL V-bottom wells, provided sterile and nest-packed for laboratory sample processing and storage. Includes a 24-well plastic comb and is suitable for automated workflows.
  • 24 wells
  • 16 mL nominal volume per well
  • V-bottom well geometry
  • Square well top for secure sealing
  • Manufactured from USP Class VI polypropylene
  • Sterile, nest-packed format
  • Includes a 24-well plastic comb
  • Suitable for storage, extraction, and automated processing