Learn More
Chemglass Life Sciences Chemglass Microplate, 384-Well Deep, 120 μL, V-bottom, Sterile
SDP
Catalog No. 502733174
Encompass_Preferred
The Chemglass Life Sciences 384-well deep well microplate offers 120 μL nominal well volume in an ANSI/SBS-compatible format suitable for automation and multichannel pipetting. Manufactured from Class VI polypropylene, the sterile plate is DNase/RNase-free and produced in a Class 100,000 clean workshop for high-throughput screening, tissue culture, and nucleic acid workflows.
  • 384-well format with 120 μL nominal well volume.
  • Deep wells with V-bottom and square well top for low residual volume.
  • Class VI polypropylene material, DNase/RNase free, non-pyrogenic.
  • Conforms to ANSI/SBS dimensions; compatible with multi-channel pipettes and automation.
  • Sterile; packaged with batch identification and produced in a Class 100,000 clean workshop.
  • Autoclavable at 121°C, 20 psi for 20 minutes.
  • Temperature stable to -80°C; centrifuge-compatible up to 3,000-4,000 g.
Catalog No. 50-273-3174 Supplier Chemglass Life Sciences Supplier No. CLS3506S381
The Chemglass Life Sciences 384-well deep well microplate offers 120 μL nominal well volume in an ANSI/SBS-compatible format suitable for automation and multichannel pipetting. Manufactured from Class VI polypropylene, the sterile plate is DNase/RNase-free and produced in a Class 100,000 clean workshop for high-throughput screening, tissue culture, and nucleic acid workflows.
  • 384-well format with 120 μL nominal well volume.
  • Deep wells with V-bottom and square well top for low residual volume.
  • Class VI polypropylene material, DNase/RNase free, non-pyrogenic.
  • Conforms to ANSI/SBS dimensions; compatible with multi-channel pipettes and automation.
  • Sterile; packaged with batch identification and produced in a Class 100,000 clean workshop.
  • Autoclavable at 121°C, 20 psi for 20 minutes.
  • Temperature stable to -80°C; centrifuge-compatible up to 3,000-4,000 g.