Learn More
Chemglass Life Sciences Chemglass Microplate, 384-Well Deep Well, 120 μL per Well
SDP
Catalog No. 502733189
Encompass_Preferred
A 384-well deep well polypropylene microplate with 120 μL well volume designed for automated workflows and multichannel pipetting. The plate conforms to ANSI/SBS dimensions, is non-sterile and DNase/RNase free, autoclavable, and suitable for storage down to -80 °C and centrifugation up to 3,000-4,000 x g.
  • 384 wells, 120 μL maximum well volume.
  • Deep well design with V-bottom and square well top for reduced residual volume.
  • Made from Class VI polypropylene; low heavy metal content.
  • ANSI/SBS footprint compatible with automation and multichannel pipettes.
  • Autoclavable; stable to -80 °C; centrifuge up to 3,000-4,000 x g.
  • Non-sterile, DNase/RNase free, non-pyrogenic.
Catalog No. 50-273-3189 Supplier Chemglass Life Sciences Supplier No. CLS3506381
Add to Cart
Add to Cart
A 384-well deep well polypropylene microplate with 120 μL well volume designed for automated workflows and multichannel pipetting. The plate conforms to ANSI/SBS dimensions, is non-sterile and DNase/RNase free, autoclavable, and suitable for storage down to -80 °C and centrifugation up to 3,000-4,000 x g.
  • 384 wells, 120 μL maximum well volume.
  • Deep well design with V-bottom and square well top for reduced residual volume.
  • Made from Class VI polypropylene; low heavy metal content.
  • ANSI/SBS footprint compatible with automation and multichannel pipettes.
  • Autoclavable; stable to -80 °C; centrifuge up to 3,000-4,000 x g.
  • Non-sterile, DNase/RNase free, non-pyrogenic.