Learn More
Chemglass Life Sciences Chemglass 384-well deep microplate, 240 μL capacity, polypropylene (non-sterile)
SDP
Catalog No. 502733188
Encompass_Preferred
384-well deep polypropylene microplate with 240 μL well capacity, V-bottom wells, and a square well top designed for high-throughput workflows and automated handling. The plate conforms to ANSI/SBS dimensions and is suitable for storage, reagent transfer, dilution, and nucleic acid workflows.
  • 384-well format with approximately 240 μL usable volume per well.
  • Made from Class VI polypropylene for chemical compatibility and stability.
  • V-bottom wells and square top for efficient pipetting and sealing.
  • Conforms to ANSI and international SBS footprint for automation compatibility.
  • Autoclavable and rated for -80 °C to 120 °C temperature range.
  • DNase/RNase free and manufactured in a clean-room environment.
  • Packaged in case quantities (pack size: CS5) for lab stocking.
Catalog No. 50-273-3188 Supplier Chemglass Life Sciences Supplier No. CLS3506382
Add to Cart
Add to Cart
384-well deep polypropylene microplate with 240 μL well capacity, V-bottom wells, and a square well top designed for high-throughput workflows and automated handling. The plate conforms to ANSI/SBS dimensions and is suitable for storage, reagent transfer, dilution, and nucleic acid workflows.
  • 384-well format with approximately 240 μL usable volume per well.
  • Made from Class VI polypropylene for chemical compatibility and stability.
  • V-bottom wells and square top for efficient pipetting and sealing.
  • Conforms to ANSI and international SBS footprint for automation compatibility.
  • Autoclavable and rated for -80 °C to 120 °C temperature range.
  • DNase/RNase free and manufactured in a clean-room environment.
  • Packaged in case quantities (pack size: CS5) for lab stocking.