Learn More
Chemglass Life Sciences Chemglass Microplate, 48-Well Deep U-Bottom, 3.5 mL, Sterile (CLS-3506-S482)
SDP
Catalog No. 502733171
Encompass_Preferred
Chemglass Life Sciences microplate, 48-well deep U-bottom plate with 3.5 mL well capacity. Made from USP Class VI polypropylene and supplied sterile in nest packaging, this plate is designed for storage, centrifugation, and automated handling.
  • 48 wells with 3.5 mL capacity per well
  • U-bottom, round well top
  • Manufactured from USP Class VI polypropylene
  • Supplied sterile in nest packaging
  • Temperature range: -80°C to 120°C
  • Autoclavable
  • Centrifugal strength up to 3,000 x g
  • Compatible with multichannel pipettes and automation
  • Non-pyrogenic, DNase/RNase-free
  • Conforms to SBS/ANSI footprint
Catalog No. 50-273-3171 Supplier Chemglass Life Sciences Supplier No. CLS3506S482
Add to Cart
Add to Cart
Chemglass Life Sciences microplate, 48-well deep U-bottom plate with 3.5 mL well capacity. Made from USP Class VI polypropylene and supplied sterile in nest packaging, this plate is designed for storage, centrifugation, and automated handling.
  • 48 wells with 3.5 mL capacity per well
  • U-bottom, round well top
  • Manufactured from USP Class VI polypropylene
  • Supplied sterile in nest packaging
  • Temperature range: -80°C to 120°C
  • Autoclavable
  • Centrifugal strength up to 3,000 x g
  • Compatible with multichannel pipettes and automation
  • Non-pyrogenic, DNase/RNase-free
  • Conforms to SBS/ANSI footprint