Learn More
Chemglass Life Sciences Microplate, 96 Deep Well, 2mL/Well, Round Well, Polypropylene, Rimless, Porvair
SDP
Catalog No. 5000980732
Encompass_Preferred
Manufactured from pre-tested polypropylene for low extractables. Alphanumeric grid-referencing. DNase/ RNase free. Packed in sealed sleeves of 5 plates. Cylindrical well with round bottom for optimal mixing and recovery. Very easy to use with automated sample handling systems. Can be stored at -80ºC. Easily sealed with matching Cap Mat or heat seal. Fits Waters AcQuity™ autosamplers. Plates are made in virgin polypropylene to minimize extractables. Maximum volume is 2. 075ml per well and working volume is a useful 1. 85ml, more than any other comparable ‘2ml Round Well’ plate in this class. The ‘common wall’ design allows the highest possible volume to be used while maintaining an overall height of just 45mm, complete with ANSI/SLAS standard base and footprint. Extra working volume is assured in a convenient round bottom, round well format.
Catalog No. 50-009-80732 Supplier Chemglass Life Sciences Supplier No. CLS3562002
Manufactured from pre-tested polypropylene for low extractables. Alphanumeric grid-referencing. DNase/ RNase free. Packed in sealed sleeves of 5 plates. Cylindrical well with round bottom for optimal mixing and recovery. Very easy to use with automated sample handling systems. Can be stored at -80ºC. Easily sealed with matching Cap Mat or heat seal. Fits Waters AcQuity™ autosamplers. Plates are made in virgin polypropylene to minimize extractables. Maximum volume is 2. 075ml per well and working volume is a useful 1. 85ml, more than any other comparable ‘2ml Round Well’ plate in this class. The ‘common wall’ design allows the highest possible volume to be used while maintaining an overall height of just 45mm, complete with ANSI/SLAS standard base and footprint. Extra working volume is assured in a convenient round bottom, round well format.